The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007006.2 | NP_008937.1 | 5 - 6 | 700 | 143 |
GenBank mRNA | AJ001810.1 | - | 5 - 6 | 553 | 143 |
AK312721.1 | - | 5 - 6 | 703 | 143 | |
BC001403.2 | - | 5 - 6 | 661 | 143 | |
BX537360.1 | - | 5 - 6 | 704 | 143 | |
CR456919.1 | - | 5 - 6 | 553 | 143 |