Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAF5
Entrez Gene ID
7188
Gene Name
TNF receptor associated factor 5
Gene Aliases
MGC:39780, RNF84
Chromosome Location
Chr.1: 211326587 - 211374946 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033910.2 NP_001029082.1 7 - 8 779 72
NM_001319207.1 NP_001306136.1 7 - 8 812 72
NM_004619.3 NP_004610.1 7 - 8 754 72
NM_145759.2 NP_665702.1 7 - 8 754 72
XM_011509957.2 XP_011508259.1 7 - 8 972 72
XM_011509959.2 XP_011508261.1 7 - 8 1803 72
XM_011509960.2 XP_011508262.1 7 - 8 815 72
XM_017002221.1 XP_016857710.1 7 - 8 2634 72
GenBank mRNA AB000509.1 - 7 - 8 748 72
AK303286.1 - 7 - 8 785 72
BC029600.1 - 7 - 8 754 72
CR536557.1 - 6 - 7 694 72
U69108.1 - 6 - 7 637 72

View Full Product Details