The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033910.2 | NP_001029082.1 | 8 - 9 | 868 | 103 |
NM_001319207.1 | NP_001306136.1 | 8 - 9 | 901 | 103 | |
NM_004619.3 | NP_004610.1 | 8 - 9 | 843 | 103 | |
NM_145759.2 | NP_665702.1 | 8 - 9 | 843 | 103 | |
XM_011509957.2 | XP_011508259.1 | 8 - 9 | 1061 | 103 | |
XM_011509959.2 | XP_011508261.1 | 8 - 9 | 1892 | 103 | |
XM_011509960.2 | XP_011508262.1 | 8 - 9 | 904 | 103 | |
XM_017002221.1 | XP_016857710.1 | 8 - 9 | 2723 | 103 | |
GenBank mRNA | AB000509.1 | - | 8 - 9 | 837 | 103 |
AK295766.1 | - | 5 - 6 | 529 | 103 | |
AK303286.1 | - | 8 - 9 | 874 | 103 | |
BC029600.1 | - | 8 - 9 | 843 | 103 | |
CR536557.1 | - | 7 - 8 | 783 | 103 | |
U69108.1 | - | 7 - 8 | 726 | 103 |