Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXO6
Entrez Gene ID
26270
Gene Name
F-box protein 6
Gene Aliases
FBG2, FBS2, FBX6, Fbx6b
Chromosome Location
Chr.1: 11664093 - 11674354 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018438.5 NP_060908.1 5 - 6 809 127
XM_005263448.4 XP_005263505.1 5 - 6 749 127
XM_005263449.4 XP_005263506.1 5 - 6 696 127
XM_005263451.4 XP_005263508.1 5 - 6 749 127
XM_006710571.3 XP_006710634.1 5 - 6 777 127
XM_011541232.2 XP_011539534.1 5 - 6 743 127
XM_017001007.1 XP_016856496.1 6 - 7 811 127
GenBank mRNA AF129536.1 - 5 - 6 778 127
AF233223.1 - 4 - 5 654 127
AK314989.1 - 5 - 6 809 127
BC020880.1 - 5 - 6 751 127

View Full Product Details