The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002864.2 | NP_002855.2 | 9 - 10 | 1012 | 68 |
XM_011520804.2 | XP_011519106.1 | 9 - 10 | 1011 | 68 | |
XM_011520805.2 | XP_011519107.1 | 9 - 10 | 1011 | 68 | |
XM_011520806.1 | XP_011519108.1 | 9 - 10 | 1011 | 68 | |
XM_011520807.2 | XP_011519109.1 | 9 - 10 | 1011 | 68 | |
XM_017019763.1 | XP_016875252.1 | 9 - 10 | 1011 | 68 | |
XM_017019764.1 | XP_016875253.1 | 9 - 10 | 1011 | 68 | |
XM_017019765.1 | XP_016875254.1 | 9 - 10 | 1011 | 68 | |
XM_017019766.1 | XP_016875255.1 | 8 - 9 | 1160 | 68 | |
XM_017019767.1 | XP_016875256.1 | 9 - 10 | 1011 | 68 | |
GenBank mRNA | AK313639.1 | - | 9 - 10 | 1011 | 68 |
BC111756.1 | - | 8 - 9 | 1160 | 68 | |
X54380.1 | - | 9 - 10 | 1012 | 68 |