The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000672.3 | NP_000663.1 | 2 - 3 | 206 | 76 |
NM_001102470.1 | NP_001095940.1 | 2 - 3 | 206 | 76 | |
GenBank mRNA | AK092768.1 | - | 2 - 3 | 206 | 76 |
BC039065.1 | - | 2 - 3 | 195 | 76 |