The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256125.1 | NP_001243054.2 | 9 - 10 | 1195 | 102 |
NM_003465.2 | NP_003456.1 | 10 - 11 | 1252 | 102 | |
GenBank mRNA | AK055165.1 | - | 9 - 10 | 1135 | 102 |
AK314331.1 | - | 10 - 11 | 1191 | 102 | |
BC069614.1 | - | 9 - 10 | 1121 | 102 | |
BC103695.1 | - | 10 - 11 | 1168 | 102 | |
BC105680.1 | - | 10 - 11 | 1168 | 102 | |
BC105681.1 | - | 10 - 11 | 1168 | 102 | |
BC105682.1 | - | 10 - 11 | 1168 | 102 | |
U29615.1 | - | 10 - 11 | 1168 | 102 |