The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270509.1 | NP_001257438.1 | 3 - 4 | 348 | 75 |
NM_003465.2 | NP_003456.1 | 3 - 4 | 348 | 75 | |
NR_045784.1 | 3 - 4 | 348 | 75 | ||
NR_045785.1 | 3 - 4 | 348 | 75 | ||
GenBank mRNA | AK131093.1 | - | 5 - 6 | 4608 | 75 |
AK225245.1 | - | 3 - 4 | 288 | 75 | |
AK309341.1 | - | 3 - 4 | 290 | 75 | |
AK314331.1 | - | 3 - 4 | 287 | 75 | |
BC069614.1 | - | 2 - 3 | 217 | 75 | |
BC103695.1 | - | 3 - 4 | 264 | 75 | |
BC105680.1 | - | 3 - 4 | 264 | 75 | |
BC105681.1 | - | 3 - 4 | 264 | 75 | |
BC105682.1 | - | 3 - 4 | 264 | 75 | |
BP304256.1 | - | 3 - 4 | 348 | 75 | |
U29615.1 | - | 3 - 4 | 264 | 75 | |
U62662.1 | - | 3 - 4 | 261 | 75 |