The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005275.3 | NP_005266.2 | 10 - 11 | 2833 | 57 |
XM_005249015.3 | XP_005249072.1 | 11 - 12 | 2296 | 57 | |
GenBank mRNA | AK026391.1 | - | 10 - 11 | 1495 | 57 |
AK293596.1 | - | 4 - 5 | 786 | 57 | |
AK298042.1 | - | 10 - 11 | 1672 | 57 | |
AK301738.1 | - | 9 - 10 | 1228 | 57 | |
AK302480.1 | - | 9 - 10 | 1561 | 57 | |
BC013959.2 | - | 10 - 11 | 1482 | 57 | |
BC018366.1 | - | 10 - 11 | 2413 | 57 | |
BC063657.1 | - | 7 - 8 | 1065 | 57 | |
BT006648.1 | - | 10 - 11 | 1442 | 57 | |
BU687484.1 | - | 2 - 3 | 57 | ||
L25665.1 | - | 10 - 11 | 1392 | 57 |