The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005275.3 | NP_005266.2 | 2 - 3 | 1623 | 85 |
XM_005249015.3 | XP_005249072.1 | 3 - 4 | 1086 | 85 | |
GenBank mRNA | AK026391.1 | - | 2 - 3 | 285 | 85 |
AK298042.1 | - | 2 - 3 | 462 | 85 | |
AK302480.1 | - | 1 - 2 | 351 | 85 | |
AK316391.1 | - | 2 - 3 | 289 | 85 | |
BC013959.2 | - | 2 - 3 | 272 | 85 | |
BC018366.1 | - | 2 - 3 | 1203 | 85 | |
BT006648.1 | - | 2 - 3 | 232 | 85 | |
L25665.1 | - | 2 - 3 | 186 | 86 |