The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005275.3 | NP_005266.2 | 5 - 6 | 1994 | 87 |
XM_005249015.3 | XP_005249072.1 | 6 - 7 | 1457 | 87 | |
GenBank mRNA | AK298042.1 | - | 5 - 6 | 833 | 87 |
AK301738.1 | - | 5 - 6 | 584 | 87 | |
AK302480.1 | - | 4 - 5 | 722 | 87 | |
AK316391.1 | - | 5 - 6 | 595 | 87 | |
BC013959.2 | - | 5 - 6 | 643 | 87 | |
BC018366.1 | - | 5 - 6 | 1574 | 87 | |
BC063657.1 | - | 2 - 3 | 226 | 87 | |
BT006648.1 | - | 5 - 6 | 603 | 87 | |
L25665.1 | - | 5 - 6 | 553 | 87 |