Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GNL1
Entrez Gene ID
2794
Gene Name
G protein nucleolar 1 (putative)
Gene Aliases
HSR1
Chromosome Location
Chr.6: 30541377 - 30557594 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005275.3 NP_005266.2 7 - 8 2294 74
XM_005249015.3 XP_005249072.1 8 - 9 1757 74
GenBank mRNA AK026391.1 - 7 - 8 956 74
AK298042.1 - 7 - 8 1133 74
AK302480.1 - 6 - 7 1022 74
AK316391.1 - 7 - 8 895 74
BC013959.2 - 7 - 8 943 74
BC018366.1 - 7 - 8 1874 74
BC063657.1 - 4 - 5 526 74
BT006648.1 - 7 - 8 903 74

View Full Product Details