The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032152.4 | NP_115528.4 | 5 - 6 | 1697 | 74 |
XM_005272502.2 | XP_005272559.1 | 5 - 6 | 1683 | 74 | |
XM_011528352.2 | XP_011526654.1 | 5 - 6 | 1703 | 74 | |
XM_011528353.2 | XP_011526655.1 | 5 - 6 | 1701 | 74 | |
GenBank mRNA | AJ272324.1 | - | 4 - 5 | 1841 | 74 |
AK308227.1 | - | 5 - 6 | 1680 | 74 | |
BC028012.1 | - | 6 - 7 | 1654 | 74 |