Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYH3
Entrez Gene ID
4621
Gene Name
myosin, heavy chain 3, skeletal muscle, embryonic
Gene Aliases
DA2A, DA2B, DA8, HEMHC, MYHC-EMB, MYHSE1, SMHCE
Chromosome Location
Chr.17: 10628526 - 10679657 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002470.3 NP_002461.2 10 - 11 989 109
XM_011523870.2 XP_011522172.1 10 - 11 2356 109
XM_011523871.2 XP_011522173.1 10 - 11 989 109
GenBank mRNA X13988.1 - 9 - 10 985 109

View Full Product Details