The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001046.2 | NP_001037.1 | 22 - 23 | 3259 | 143 |
NM_001256461.1 | NP_001243390.1 | 21 - 22 | 3211 | 143 | |
NR_046207.1 | 22 - 23 | 3259 | 143 | ||
XM_017009771.1 | XP_016865260.1 | 21 - 22 | 2409 | 143 | |
GenBank mRNA | AF439152.1 | - | 8 - 9 | 832 | 143 |
AY280459.1 | - | 22 - 23 | 3284 | 143 | |
BC144221.1 | - | 21 - 22 | 3154 | 143 | |
BC146839.1 | - | 21 - 22 | 3154 | 143 | |
U30246.1 | - | 22 - 23 | 3259 | 143 |