Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD12
Entrez Gene ID
23253
Gene Name
ankyrin repeat domain 12
Gene Aliases
ANCO-2, ANCO1, GAC-1, Nbla00144
Chromosome Location
Chr.18: 9136753 - 9285985 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083625.2 NP_001077094.1 4 - 5 641 75
NM_001204056.1 NP_001190985.1 4 - 5 739 75
NM_015208.4 NP_056023.3 5 - 6 710 75
XM_005258092.3 XP_005258149.1 5 - 6 843 75
XM_005258093.3 XP_005258150.1 5 - 6 827 75
XM_011525638.2 XP_011523940.1 5 - 6 682 75
XM_017025663.1 XP_016881152.1 4 - 5 726 75
XM_017025667.1 XP_016881156.1 5 - 6 624 75
GenBank mRNA AF317425.1 - 4 - 5 446 75
AK024808.1 - 5 - 6 660 75
AL834204.1 - 4 - 5 569 75
AY373757.1 - 5 - 6 660 75
AY533564.2 - 4 - 5 739 75
BC057225.1 - 5 - 6 659 75
BC070234.1 - 4 - 5 611 75

View Full Product Details