Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SNHG7
- Entrez Gene ID
-
84973
- Gene Name
- small nucleolar RNA host gene 7
- Gene Aliases
-
NCRNA00061
- Chromosome Location
-
Chr.9: 136724594 - 136728184 on Build GRCh38
- Gene Symbol
- SNORA17B
- Entrez Gene ID
-
677824
- Gene Name
- small nucleolar RNA, H/ACA box 17B
- Gene Aliases
-
ACA43, SNORA43
- Chromosome Location
-
Chr.9: 136726104 - 136726237 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK054908.1 |
- |
1 - 2 |
305 |
68 |
View Full Product Details