The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303279.1 | NP_001290208.1 | 12 - 13 | 1833 | 144 |
NM_015194.2 | NP_056009.1 | 12 - 13 | 1833 | 144 | |
XM_017024685.1 | XP_016880174.1 | 12 - 13 | 1804 | 144 | |
GenBank mRNA | AB018270.2 | - | 12 - 13 | 1544 | 144 |
AK026920.1 | - | 9 - 10 | 1129 | 144 | |
AK127942.1 | - | 12 - 13 | 1804 | 144 | |
BC146763.1 | - | 12 - 13 | 1544 | 144 | |
BX537615.1 | - | 11 - 12 | 1267 | 144 |