The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303279.1 | NP_001290208.1 | 16 - 17 | 2426 | 91 |
NM_015194.2 | NP_056009.1 | 16 - 17 | 2426 | 91 | |
XM_017024685.1 | XP_016880174.1 | 16 - 17 | 2397 | 91 | |
GenBank mRNA | AB018270.2 | - | 16 - 17 | 2137 | 91 |
AK026920.1 | - | 13 - 14 | 1722 | 91 | |
AK127942.1 | - | 16 - 17 | 2395 | 91 | |
BC146763.1 | - | 16 - 17 | 2137 | 91 | |
BX537615.1 | - | 15 - 16 | 1860 | 91 |