Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOS2
Entrez Gene ID
4843
Gene Name
nitric oxide synthase 2
Gene Aliases
HEP-NOS, INOS, NOS, NOS2A
Chromosome Location
Chr.17: 27756766 - 27804339 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000625.4 NP_000616.3 12 - 13 1739 61
XM_011524859.2 XP_011523161.1 12 - 13 1765 61
XM_011524860.2 XP_011523162.1 11 - 12 1558 61
XM_011524861.1 XP_011523163.1 12 - 13 1737 61
XM_011524862.1 XP_011523164.1 10 - 11 1336 61
GenBank mRNA AB022318.1 - 10 - 11 1358 61
AF068236.1 - 12 - 13 1709 61
AK304252.1 - 12 - 13 1737 61
BC130283.1 - 12 - 13 1659 61
BC144126.1 - 12 - 13 1659 61
D26525.1 - 11 - 12 1489 61
L09210.1 - 12 - 13 1681 61
L24553.1 - 11 - 12 1551 61
U05810.1 - 12 - 13 1669 61
U20141.1 - 11 - 12 1475 61
U31511.1 - 12 - 13 1580 61
X73029.1 - 12 - 13 1700 61

View Full Product Details