The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000625.4 | NP_000616.3 | 19 - 20 | 2512 | 70 |
XM_011524859.2 | XP_011523161.1 | 19 - 20 | 2538 | 70 | |
XM_011524860.2 | XP_011523162.1 | 18 - 19 | 2331 | 70 | |
XM_011524861.1 | XP_011523163.1 | 19 - 20 | 2510 | 70 | |
XM_011524862.1 | XP_011523164.1 | 17 - 18 | 2109 | 70 | |
GenBank mRNA | AB022318.1 | - | 17 - 18 | 2131 | 70 |
AF068236.1 | - | 19 - 20 | 2482 | 70 | |
BC130283.1 | - | 19 - 20 | 2432 | 70 | |
BC144126.1 | - | 19 - 20 | 2432 | 70 | |
D26525.1 | - | 18 - 19 | 2262 | 70 | |
L09210.1 | - | 19 - 20 | 2454 | 70 | |
L24553.1 | - | 18 - 19 | 2324 | 70 | |
U05810.1 | - | 19 - 20 | 2442 | 70 | |
U20141.1 | - | 18 - 19 | 2248 | 70 | |
U31511.1 | - | 19 - 20 | 2353 | 70 | |
X73029.1 | - | 19 - 20 | 2473 | 70 |