The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127384.2 | NP_001120856.1 | 2 - 3 | 248 | 107 |
NM_001291133.1 | NP_001278062.1 | 3 - 4 | 325 | 107 | |
NM_013266.3 | NP_037398.2 | 2 - 3 | 295 | 107 | |
XM_017016151.1 | XP_016871640.1 | 2 - 3 | 221 | 107 | |
XM_017016152.1 | XP_016871641.1 | 3 - 4 | 398 | 107 | |
XM_017016153.1 | XP_016871642.1 | 3 - 4 | 298 | 107 | |
XM_017016158.1 | XP_016871647.1 | 3 - 4 | 396 | 107 | |
GenBank mRNA | AF091606.1 | - | 2 - 3 | 273 | 107 |
AK289756.1 | - | 2 - 3 | 184 | 107 | |
AK308726.1 | - | 2 - 3 | 162 | 107 | |
AK313882.1 | - | 2 - 3 | 298 | 107 | |
AY198183.1 | - | 3 - 4 | 264 | 107 | |
BC022004.2 | - | 3 - 4 | 320 | 107 | |
BC030646.2 | - | 3 - 4 | 320 | 107 | |
BC065819.1 | - | 2 - 3 | 191 | 107 | |
BP362015.1 | - | 2 - 3 | 221 | 107 |