The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031471.5 | NP_113659.3 | 10 - 11 | 1355 | 62 |
NM_178443.2 | NP_848537.1 | 10 - 11 | 1367 | 62 | |
XM_011545294.2 | XP_011543596.1 | 10 - 11 | 1379 | 62 | |
XM_011545295.2 | XP_011543597.1 | 9 - 10 | 1212 | 62 | |
XM_017018398.1 | XP_016873887.1 | 10 - 11 | 1363 | 62 | |
XM_017018399.1 | XP_016873888.1 | 9 - 10 | 1215 | 62 | |
GenBank mRNA | AK093719.1 | - | 10 - 11 | 1300 | 62 |
AY093951.1 | - | 10 - 11 | 1301 | 62 | |
AY093952.1 | - | 10 - 11 | 1313 | 62 | |
BC004347.1 | - | 10 - 11 | 1299 | 62 | |
BC013366.2 | - | 10 - 11 | 1299 | 62 | |
BC015584.2 | - | 10 - 11 | 1294 | 62 |