The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171976.1 | NP_001165447.1 | 11 - 12 | 1239 | 69 |
NM_017675.4 | NP_060145.3 | 11 - 12 | 1053 | 69 | |
GenBank mRNA | AB047004.1 | - | 10 - 11 | 1003 | 69 |
AK000390.1 | - | 6 - 7 | 608 | 69 | |
AK315055.1 | - | 11 - 12 | 1239 | 69 | |
BC130282.1 | - | 10 - 11 | 1012 | 69 |