The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171976.1 | NP_001165447.1 | 20 - 21 | 3082 | 112 |
NM_017675.4 | NP_060145.3 | 20 - 21 | 2896 | 112 | |
GenBank mRNA | AB047004.1 | - | 19 - 20 | 2846 | 112 |
AK000131.1 | - | 5 - 6 | 1035 | 112 | |
AK000390.1 | - | 15 - 16 | 2450 | 112 | |
AK225217.1 | - | 5 - 6 | 1035 | 112 | |
AK315055.1 | - | 20 - 21 | 3082 | 112 | |
BC130282.1 | - | 19 - 20 | 2855 | 112 |