The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171976.1 | NP_001165447.1 | 7 - 8 | 764 | 55 |
NM_017675.4 | NP_060145.3 | 7 - 8 | 578 | 55 | |
GenBank mRNA | AB047004.1 | - | 6 - 7 | 528 | 55 |
AK000390.1 | - | 2 - 3 | 133 | 55 | |
AK315055.1 | - | 7 - 8 | 764 | 55 | |
BC130282.1 | - | 6 - 7 | 537 | 55 | |
BX478954.1 | - | 7 - 8 | 565 | 55 |