The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004457.3 | NP_004448.2 | 1 - 2 | 243 | 134 |
NM_203372.1 | NP_976251.1 | 1 - 2 | 243 | 134 | |
GenBank mRNA | AB061712.1 | - | 1 - 2 | 211 | 134 |
AK314621.1 | - | 1 - 2 | 221 | 134 | |
BC041692.1 | - | 1 - 2 | 235 | 134 | |
BG772246.1 | - | 1 - 2 | 328 | 134 | |
BX472576.1 | - | 1 - 2 | 237 | 134 |