The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193455.1 | NP_001180384.1 | 12 - 13 | 1783 | 153 |
NM_017755.5 | NP_060225.4 | 13 - 14 | 1888 | 153 | |
NR_037947.1 | 12 - 13 | 1803 | 153 | ||
GenBank mRNA | AB255451.1 | - | - | 1887 | 153 |
AK000310.1 | - | 7 - 8 | 884 | 153 | |
AK023994.1 | - | 1 - 2 | 154 | 153 | |
AK291144.1 | - | 13 - 14 | 1571 | 153 | |
AK298980.1 | - | 12 - 13 | 1465 | 153 | |
BC001041.2 | - | 11 - 12 | 1244 | 153 | |
BC137083.1 | - | 13 - 14 | 1569 | 153 |