The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019592.6 | NP_062538.5 | 10 - 11 | 1365 | 95 |
XM_011518862.1 | XP_011517164.1 | 10 - 11 | 1642 | 95 | |
GenBank mRNA | AF265230.1 | - | 10 - 11 | 1363 | 95 |
AK000389.1 | - | 9 - 10 | 1256 | 95 | |
AK000697.1 | - | 9 - 10 | 1256 | 95 | |
AK022300.1 | - | 5 - 6 | 637 | 95 | |
AK022532.1 | - | 10 - 11 | 1361 | 95 | |
AK314401.1 | - | 10 - 11 | 1320 | 95 | |
BC016691.1 | - | 11 - 12 | 1320 | 95 | |
BC032618.1 | - | 6 - 7 | 688 | 95 | |
BC063115.1 | - | 10 - 11 | 1346 | 95 | |
BC110584.1 | - | 10 - 11 | 1358 | 95 | |
BC110585.1 | - | 10 - 11 | 1358 | 95 | |
BC152309.1 | - | 10 - 11 | 1358 | 95 |