Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C2orf42
Entrez Gene ID
54980
Gene Name
chromosome 2 open reading frame 42
Gene Aliases
-
Chromosome Location
Chr.2: 70149885 - 70191071 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005264390.2 XP_005264447.1 1 - 2 101 97

View Full Product Details