Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C2orf42
Entrez Gene ID
54980
Gene Name
chromosome 2 open reading frame 42
Gene Aliases
-
Chromosome Location
Chr.2: 70149885 - 70191071 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017880.1 NP_060350.1 4 - 5 1260 89
XM_005264389.2 XP_005264446.1 4 - 5 1453 89
XM_005264390.2 XP_005264447.1 4 - 5 1322 89
XM_005264391.2 XP_005264448.1 4 - 5 1404 89
XM_011532937.2 XP_011531239.1 3 - 4 1198 89
XM_017004398.1 XP_016859887.1 4 - 5 1741 89
XM_017004399.1 XP_016859888.1 4 - 5 1449 89
XM_017004400.1 XP_016859889.1 4 - 5 1333 89
GenBank mRNA AK000565.1 - 4 - 5 1260 89
AK026303.1 - 2 - 3 545 89
AL136834.1 - 3 - 4 1038 89
BC002825.2 - 4 - 5 1249 89
BC005079.1 - 4 - 5 1249 89

View Full Product Details