Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KANSL1
Entrez Gene ID
284058
Gene Name
KAT8 regulatory NSL complex subunit 1
Gene Aliases
CENP-36, KDVS, KIAA1267, MSL1v1, NSL1, hMSL1v1
Chromosome Location
Chr.17: 46029916 - 46225374 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193465.1 NP_001180394.1 10 - 11 2649 84
NM_001193466.1 NP_001180395.1 9 - 10 2863 84
NM_015443.3 NP_056258.1 9 - 10 2565 84
XM_006721823.1 XP_006721886.1 10 - 11 2649 84
XM_006721824.3 XP_006721887.1 10 - 11 2833 84
XM_011524628.2 XP_011522930.1 10 - 11 2833 84
XM_011524629.2 XP_011522931.1 9 - 10 2731 84
XM_011524632.2 XP_011522934.1 9 - 10 1259 84
XM_017024489.1 XP_016879978.1 8 - 9 2658 84
GenBank mRNA AB033093.1 - 8 - 9 2486 84
AK291259.1 - 9 - 10 2626 84
AL137317.1 - 5 - 6 807 84
BC035892.1 - 4 - 5 670 84
BC098376.1 - 9 - 10 2565 84
BX538006.1 - 9 - 10 2866 84
BX648760.1 - 2 - 3 6623 84

View Full Product Details