Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IQCB1
Entrez Gene ID
9657
Gene Name
IQ motif containing B1
Gene Aliases
NPHP5, PIQ, SLSN5
Chromosome Location
Chr.3: 121769761 - 121835079 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001023570.3 NP_001018864.2 13 - 14 1625 74
NM_001023571.3 NP_001018865.2 10 - 11 1226 74
NM_001319107.1 NP_001306036.1 13 - 14 1624 74
NR_134968.1 12 - 13 1514 74
XM_005247912.2 XP_005247969.1 12 - 13 1438 74
XM_011513335.2 XP_011511637.1 11 - 12 1344 74
XM_017007537.1 XP_016863026.1 12 - 13 1507 74
XM_017007538.1 XP_016863027.1 9 - 10 1309 74
GenBank mRNA AB062481.1 - 12 - 13 1508 74
AY714228.1 - 13 - 14 1619 74
AY964667.1 - 10 - 11 1176 74
AY964668.1 - 13 - 14 1563 74
BC005806.2 - 10 - 11 1167 74
D25278.1 - 13 - 14 1566 74

View Full Product Details