Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABR
Entrez Gene ID
29
Gene Name
active BCR-related
Gene Aliases
MDB
Chromosome Location
Chr.17: 1003518 - 1229713 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001092.4 NP_001083.2 13 - 14 1483 62
NM_001159746.2 NP_001153218.1 14 - 15 2024 62
NM_001282149.1 NP_001269078.1 9 - 10 1195 62
NM_001322840.1 NP_001309769.1 14 - 15 1727 62
NM_001322841.1 NP_001309770.1 14 - 15 2389 62
NM_021962.4 NP_068781.2 14 - 15 1809 62
XM_011523810.2 XP_011522112.1 14 - 15 1612 62
XM_011523811.2 XP_011522113.2 14 - 15 2422 62
XM_011523812.2 XP_011522114.1 13 - 14 2959 62
XM_011523813.2 XP_011522115.1 13 - 14 1483 62
XM_011523814.2 XP_011522116.1 14 - 15 2024 62
XM_011523815.2 XP_011522117.1 14 - 15 1707 62
XM_017024539.1 XP_016880028.1 14 - 15 2419 62
XM_017024540.1 XP_016880029.1 13 - 14 2321 62
XM_017024541.1 XP_016880030.1 14 - 15 2422 62
XM_017024542.1 XP_016880031.1 9 - 10 1180 62
XM_017024543.1 XP_016880032.1 9 - 10 1190 62
GenBank mRNA AK124547.1 - 14 - 15 62
AK295191.1 - 13 - 14 1401 62
AK299909.1 - 9 - 10 1050 62
AK302682.1 - 14 - 15 1550 62
CF135918.1 - 5 - 6 530 62
U01147.1 - 14 - 15 1672 62

View Full Product Details