The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321881.1 | NP_001308810.1 | 14 - 15 | 1360 | 66 |
NM_001321882.1 | NP_001308811.1 | 14 - 15 | 1578 | 66 | |
NM_024635.3 | NP_078911.3 | 14 - 15 | 1364 | 66 | |
XM_005252127.3 | XP_005252184.1 | 14 - 15 | 1435 | 66 | |
XM_011518903.2 | XP_011517205.1 | 11 - 12 | 1167 | 66 | |
XM_011518904.2 | XP_011517206.1 | 8 - 9 | 853 | 66 | |
XM_017015012.1 | XP_016870501.1 | 15 - 16 | 1423 | 66 | |
GenBank mRNA | AK025266.1 | - | 14 - 15 | 1360 | 66 |
AK225237.1 | - | 14 - 15 | 1360 | 66 | |
AK292239.1 | - | 14 - 15 | 1364 | 66 | |
BC117427.1 | - | 14 - 15 | 1285 | 66 | |
BC117429.1 | - | 14 - 15 | 1281 | 66 |