The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321881.1 | NP_001308810.1 | 17 - 18 | 1704 | 102 |
NM_001321882.1 | NP_001308811.1 | 17 - 18 | 1922 | 102 | |
NM_024635.3 | NP_078911.3 | 17 - 18 | 1708 | 102 | |
XM_005252127.3 | XP_005252184.1 | 17 - 18 | 1779 | 102 | |
XM_011518903.2 | XP_011517205.1 | 14 - 15 | 1511 | 102 | |
XM_011518904.2 | XP_011517206.1 | 11 - 12 | 1197 | 102 | |
XM_017015012.1 | XP_016870501.1 | 18 - 19 | 1767 | 102 | |
GenBank mRNA | AK025266.1 | - | 17 - 18 | 1705 | 103 |
AK225237.1 | - | 17 - 18 | 1705 | 103 | |
BC117427.1 | - | 17 - 18 | 1629 | 102 | |
BC117429.1 | - | 17 - 18 | 1625 | 102 |