The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000321.2 | NP_000312.2 | 25 - 26 | 2824 | 117 |
XM_011535171.2 | XP_011533473.1 | 24 - 25 | 2413 | 117 | |
GenBank mRNA | AB208788.1 | - | 25 - 26 | 2755 | 117 |
AK291258.1 | - | 25 - 26 | 2845 | 117 | |
AK299179.1 | - | 26 - 27 | 2735 | 117 | |
AK300284.1 | - | 26 - 27 | 2761 | 117 | |
AK307125.1 | - | 1 - 2 | 379 | 117 | |
BC039060.1 | - | 25 - 26 | 2810 | 117 | |
BC040540.1 | - | 25 - 26 | 2820 | 117 | |
L41870.1 | - | 25 - 26 | 2795 | 117 | |
M15400.1 | - | 25 - 26 | 2796 | 117 | |
M28419.1 | - | 25 - 26 | 2796 | 117 | |
M33647.1 | - | 25 - 26 | 2661 | 117 |