Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RB1
Entrez Gene ID
5925
Gene Name
RB transcriptional corepressor 1
Gene Aliases
OSRC, PPP1R130, RB, p105-Rb, pRb, pp110
Chromosome Location
Chr.13: 48303747 - 48481890 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000321.2 NP_000312.2 2 - 3 426 93
GenBank mRNA AK291258.1 - 2 - 3 93
AK299179.1 - 3 - 4 337 93
AK300284.1 - 3 - 4 363 93
AK307730.1 - 2 - 3 413 93
BC039060.1 - 2 - 3 412 93
BC040540.1 - 2 - 3 422 93
L41870.1 - 2 - 3 397 93
M15400.1 - 2 - 3 398 93
M28419.1 - 2 - 3 398 93
M33647.1 - 2 - 3 263 93

View Full Product Details