Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDR45
Entrez Gene ID
11152
Gene Name
WD repeat domain 45
Gene Aliases
JM5, NBIA4, NBIA5, WDRX1, WIPI-4, WIPI4
Chromosome Location
Chr.X: 49074433 - 49101121 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029896.1 NP_001025067.1 9 - 10 1005 68
NM_007075.3 NP_009006.2 10 - 11 1275 68
GenBank mRNA AJ005897.1 - 9 - 10 961 68
AK023344.1 - 7 - 8 1983 68
AK225122.1 - 9 - 10 984 68
AK315717.1 - 9 - 10 992 68
AY691428.1 - 8 - 9 833 68
BC000464.2 - 9 - 10 999 68
BC003037.1 - 9 - 10 999 68
BC009027.1 - 6 - 7 662 68
BC069206.1 - 10 - 11 1275 68
CR456725.1 - 8 - 9 833 68

View Full Product Details