Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDR45
Entrez Gene ID
11152
Gene Name
WD repeat domain 45
Gene Aliases
JM5, NBIA4, NBIA5, WDRX1, WIPI-4, WIPI4
Chromosome Location
Chr.X: 49074433 - 49101121 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029896.1 NP_001025067.1 10 - 11 1147 69
NM_007075.3 NP_009006.2 11 - 12 1417 69
GenBank mRNA AJ005897.1 - 10 - 11 1103 69
AK225122.1 - 10 - 11 1126 69
AY691428.1 - 9 - 10 975 69
BC000464.2 - 10 - 11 1141 69
BC003037.1 - 10 - 11 1141 69
BC009027.1 - 7 - 8 804 69
BC069206.1 - 11 - 12 1417 69
CR456725.1 - 9 - 10 975 69

View Full Product Details