The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029896.1 | NP_001025067.1 | 3 - 4 | 302 | 66 |
NM_007075.3 | NP_009006.2 | 4 - 5 | 569 | 66 | |
GenBank mRNA | AJ005897.1 | - | 3 - 4 | 258 | 66 |
AK225122.1 | - | 3 - 4 | 281 | 66 | |
AK301084.1 | - | 3 - 4 | 283 | 66 | |
AK315717.1 | - | 3 - 4 | 286 | 66 | |
AY691428.1 | - | 2 - 3 | 130 | 66 | |
BC000464.2 | - | 3 - 4 | 296 | 66 | |
BC003037.1 | - | 3 - 4 | 296 | 66 | |
BC069206.1 | - | 4 - 5 | 569 | 66 | |
BQ068083.1 | - | 3 - 4 | 302 | 66 | |
CR456725.1 | - | 2 - 3 | 130 | 66 |