The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142293.1 | NP_001135765.1 | 12 - 13 | 1408 | 126 |
NM_014570.4 | NP_055385.3 | 13 - 14 | 1540 | 126 | |
XM_005261525.3 | XP_005261582.1 | 12 - 13 | 1298 | 126 | |
GenBank mRNA | AF111847.1 | - | 13 - 14 | 1377 | 126 |
AK002083.1 | - | 10 - 11 | 969 | 126 | |
AK297635.1 | - | 11 - 12 | 1190 | 126 | |
AK300531.1 | - | 12 - 13 | 1181 | 126 | |
AK307967.1 | - | 12 - 13 | 1250 | 126 | |
AK315720.1 | - | 13 - 14 | 1378 | 126 | |
AL137598.1 | - | 12 - 13 | 1192 | 126 | |
AL159143.1 | - | 13 - 14 | 1386 | 126 | |
BC005122.2 | - | 13 - 14 | 1349 | 126 | |
CR456382.1 | - | 13 - 14 | 1331 | 126 |