The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003811.1 | NP_001003811.1 | 30 - 31 | 2894 | 93 |
NM_031276.2 | NP_112566.2 | 29 - 30 | 2772 | 93 | |
XM_011530996.1 | XP_011529298.1 | 18 - 19 | 1912 | 93 | |
GenBank mRNA | AF285594.1 | - | 28 - 29 | 2677 | 93 |
AK057391.1 | - | 18 - 19 | 1913 | 93 | |
AK057523.1 | - | 30 - 31 | 2888 | 93 | |
AK292471.1 | - | 29 - 30 | 2766 | 93 | |
BC036016.1 | - | 29 - 30 | 2772 | 93 |