The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003811.1 | NP_001003811.1 | 4 - 5 | 366 | 71 |
NM_031276.2 | NP_112566.2 | 3 - 4 | 244 | 71 | |
XM_011530994.1 | XP_011529296.1 | 3 - 4 | 238 | 71 | |
XM_017029649.1 | XP_016885138.1 | 3 - 4 | 238 | 71 | |
GenBank mRNA | AF285594.1 | - | 2 - 3 | 149 | 71 |
AK292471.1 | - | 3 - 4 | 238 | 71 | |
BC036016.1 | - | 3 - 4 | 244 | 71 |