The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319072.1 | NP_001306001.1 | 4 - 5 | 586 | 100 |
NM_001319073.1 | NP_001306002.1 | 6 - 7 | 754 | 100 | |
NM_014016.4 | NP_054735.3 | 5 - 6 | 714 | 100 | |
XM_011533500.2 | XP_011531802.1 | 5 - 6 | 505 | 100 | |
GenBank mRNA | AB020658.1 | - | 5 - 6 | 558 | 100 |
AK291142.1 | - | 5 - 6 | 598 | 100 | |
AK296421.1 | - | 4 - 5 | 368 | 100 | |
AK301248.1 | - | 6 - 7 | 594 | 100 | |
AL136831.1 | - | 5 - 6 | 504 | 100 | |
BC016559.1 | - | 5 - 6 | 510 | 100 | |
CR749564.1 | - | 5 - 6 | 1660 | 100 |