The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001279353.1 | NP_001266282.1 | 3 - 4 | 517 | 86 |
NM_001279354.1 | NP_001266283.1 | 3 - 4 | 506 | 86 | |
NM_007259.4 | NP_009190.2 | 3 - 4 | 861 | 86 | |
NR_103998.1 | 3 - 4 | 486 | 86 | ||
XM_017000164.1 | XP_016855653.1 | 3 - 4 | 411 | 86 | |
GenBank mRNA | AF165513.1 | - | 3 - 4 | 341 | 86 |
AJ133421.1 | - | 3 - 4 | 329 | 86 | |
AK023170.1 | - | 3 - 4 | 348 | 86 | |
AK223214.1 | - | 3 - 4 | 314 | 86 | |
AK293555.1 | - | 3 - 4 | 347 | 86 | |
AK295529.1 | - | 3 - 4 | 361 | 86 | |
AK298786.1 | - | 3 - 4 | 428 | 86 | |
AK301994.1 | - | 3 - 4 | 468 | 86 | |
AK307949.1 | - | 3 - 4 | 307 | 86 | |
BC012932.1 | - | 3 - 4 | 347 | 86 | |
BC028382.2 | - | 3 - 4 | 842 | 86 | |
BG176899.1 | - | 3 - 4 | 516 | 86 | |
DA374440.1 | - | 3 - 4 | 486 | 86 |