The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284297.1 | NP_001271226.1 | 4 - 5 | 674 | 73 |
NM_001284298.1 | NP_001271227.1 | 4 - 5 | 674 | 73 | |
NM_001284299.1 | NP_001271228.1 | 4 - 5 | 674 | 73 | |
NM_004573.2 | NP_004564.2 | 4 - 5 | 674 | 73 | |
XM_011521674.2 | XP_011519976.1 | 4 - 5 | 649 | 73 | |
XM_017022314.1 | XP_016877803.1 | 4 - 5 | 649 | 73 | |
XM_017022315.1 | XP_016877804.1 | 4 - 5 | 649 | 73 | |
XM_017022316.1 | XP_016877805.1 | 4 - 5 | 649 | 73 | |
XM_017022317.1 | XP_016877806.1 | 4 - 5 | 649 | 73 | |
XM_017022318.1 | XP_016877807.1 | 4 - 5 | 649 | 73 | |
XM_017022319.1 | XP_016877808.1 | 4 - 5 | 649 | 73 | |
GenBank mRNA | AB209583.1 | - | 4 - 5 | 394 | 73 |
BC000939.3 | - | 4 - 5 | 489 | 73 | |
BC009009.1 | - | 4 - 5 | 611 | 73 | |
BC136467.1 | - | 4 - 5 | 526 | 73 | |
BT006905.1 | - | 4 - 5 | 373 | 73 | |
M95678.1 | - | 4 - 5 | 537 | 73 |