The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001560.2 | NP_001001560.1 | 10 - 11 | 1291 | 120 |
NM_001172687.1 | NP_001166158.1 | 12 - 13 | 1552 | 120 | |
NM_001172688.1 | NP_001166159.1 | 13 - 14 | 1598 | 120 | |
NM_013365.4 | NP_037497.1 | 12 - 13 | 1552 | 120 | |
XM_005261517.3 | XP_005261574.1 | 13 - 14 | 1777 | 120 | |
XM_005261518.1 | XP_005261575.1 | 12 - 13 | 1237 | 120 | |
XM_005261520.1 | XP_005261577.1 | 12 - 13 | 1443 | 120 | |
XM_005261522.1 | XP_005261579.1 | 7 - 8 | 794 | 120 | |
XM_006724229.1 | XP_006724292.1 | 13 - 14 | 1485 | 120 | |
XM_011530122.1 | XP_011528424.1 | 13 - 14 | 1710 | 120 | |
XM_011530123.1 | XP_011528425.1 | 12 - 13 | 1288 | 120 | |
XM_011530124.1 | XP_011528426.1 | 11 - 12 | 1516 | 120 | |
XM_017028760.1 | XP_016884249.1 | 13 - 14 | 1726 | 120 | |
XM_017028761.1 | XP_016884250.1 | 11 - 12 | 2893 | 120 | |
XM_017028762.1 | XP_016884251.1 | 11 - 12 | 1465 | 120 | |
XM_017028763.1 | XP_016884252.1 | 13 - 14 | 1412 | 120 | |
XM_017028764.1 | XP_016884253.1 | 10 - 11 | 1182 | 120 | |
XM_017028765.1 | XP_016884254.1 | 6 - 7 | 727 | 120 | |
GenBank mRNA | AF190862.1 | - | 12 - 13 | 1180 | 120 |
AF218584.1 | - | 12 - 13 | 1169 | 120 | |
AF233521.1 | - | 12 - 13 | 1187 | 120 | |
AK075256.1 | - | 12 - 13 | 1530 | 120 | |
AK095427.1 | - | 5 - 6 | 442 | 120 | |
AK122898.1 | - | 13 - 14 | 1598 | 120 | |
AK225157.1 | - | 12 - 13 | 1186 | 120 | |
AK290548.1 | - | 12 - 13 | 1191 | 120 | |
AL110219.1 | - | 11 - 12 | 1076 | 120 | |
BC000538.2 | - | 3 - 4 | 345 | 120 | |
BC044629.1 | - | 10 - 11 | 920 | 120 | |
CR456493.1 | - | 12 - 13 | 1352 | 120 |