Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SREBF2
Entrez Gene ID
6721
Gene Name
sterol regulatory element binding transcription factor 2
Gene Aliases
SREBP-2, SREBP2, bHLHd2
Chromosome Location
Chr.22: 41833079 - 41907308 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004599.3 NP_004590.2 6 - 7 1392 64
NR_103834.1 6 - 7 1392 64
XM_006724310.2 XP_006724373.1 6 - 7 1636 64
XM_011530347.1 XP_011528649.1 6 - 7 1221 64
XM_017028921.1 XP_016884410.1 6 - 7 1221 64
XM_017028922.1 XP_016884411.1 6 - 7 1221 64
GenBank mRNA BC051385.1 - 6 - 7 1298 64
BC051799.1 - 6 - 7 1358 64
BC056158.1 - 6 - 7 1338 64
CT841522.1 - 6 - 7 1282 64
U02031.1 - 6 - 7 1317 64

View Full Product Details