The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165036.1 | NP_001158508.1 | 16 - 17 | 118 | |
NM_002541.3 | NP_002532.2 | 16 - 17 | 118 | ||
XM_005249759.4 | XP_005249816.1 | 17 - 18 | 118 | ||
XM_005249761.3 | XP_005249818.1 | 17 - 18 | 118 | ||
XM_011515408.2 | XP_011513710.1 | 18 - 19 | 118 | ||
GenBank mRNA | AK293871.1 | - | 16 - 17 | 118 | |
AK294951.1 | - | 13 - 14 | 118 | ||
AK296400.1 | - | 13 - 14 | 118 | ||
AK302805.1 | - | 14 - 15 | 118 | ||
AK304439.1 | - | 16 - 17 | 118 | ||
AK304690.1 | - | 14 - 15 | 118 | ||
AK314179.1 | - | 16 - 17 | 118 | ||
BC004964.1 | - | 15 - 16 | 118 | ||
BC014617.2 | - | 15 - 16 | 118 | ||
D10523.1 | - | 16 - 17 | 2234 | 118 |